Learn More
Melford X-GlcA Sodium Salt
Product Code. 30018102
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
500mg
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018102 5 g 5g
30018100 500 mg 500mg
30018099 250 mg 250mg
30018098 100 mg 100mg
30018101 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30018102 Supplier Melford Supplier No. B721005.0
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
Histochemical substrate for β-D-Glucuronidase.
Histochemical substrate for β-D-Glucuronidase (GUS) encoded by the gusA gene. The substrate is used as a qualitative histochemical marker of specific GUS expression in cells and tissue. X-GlcA is cleaved by GUS at the B1 glucuronic bond between glucuronic acid and the 5-Bromo-4-chloro-3-indolyl part of X-GlcA via hydrolysis. The cleavage of X-GlcA results in the precipitation of water insoluble blue dichloro-dibromo-indigo precipitate at the site of enzymatic cleavage. Allow to warm to room temperature before using.
Specifications
Quantity 5 g