Learn More
ABEBA™ X-Light Black/Yellow Safety Trainers S3 ESD
Product Code. 17161565
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17161565 35 1 pair
17171565 36 1 pair
17161575 45 1 pair
17171575 46 1 pair
17181575 47 1 pair
17191575 48 1 pair
17181565 37 1 pair
17191565 38 1 pair
17101575 39 1 pair
17111575 40 1 pair
17121575 41 1 pair
17131575 42 1 pair
17141575 43 1 pair
17151575 44 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17161565 Supplier ABEBA™ Supplier No. 7131860/35
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Black/yellow X-light safety trainer with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, textile antipuncture midsole, PU slip resistant outsole
  • Lightweight, comfortable occupational shoes
  • Comply to EN ISO 20345: 2012, S3 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant Mono PU outsole SRC
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Black and yellow Cordura
  • Silver ions lining
  • Replaceable active comfort insole
Specifications
Color Black, Yellow
Material Cordura
Size 35
Toe Material Steel
Certifications/Compliance (Europe) Comply to EN 20345: 2012 S3 SRC, ESD according EN 61340
Gender Unisex
Ankle Style Trainers
Waterproof Water Resistant
Product Type S3 SRC ESD