Learn More
Abeba™ X-Light Safety Loafer
Product Code. 17692035
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17692035 42 1 pair
17622035 35 1 pair
17632035 36 1 pair
17642035 37 1 pair
17652035 38 1 pair
17662035 39 1 pair
17672035 40 1 pair
17682035 41 1 pair
17602045 43 1 pair
17612045 44 1 pair
17622045 45 1 pair
17632045 46 1 pair
17642045 47 1 pair
17652045 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17692035 Supplier Abeba™ Supplier No. 711032/42
  • ONLINE EXCLUSIVE PRICE
    140.00€
    76.89€ / 1 pairs
    Save 63.11€
    45% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Abeba X-Light White Safety Loafer - Smooth leather safety shoes with elastic instep.
  • CE, EN ISO 20345:2011, S2, SRC
  • Smooth leather
  • Black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable Active Comfort Insole (art. 351620)
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with elastic band
Specifications
Closure Type Elastic Band
Color White
Material Leather
Size 42
Toe Material Steel
Certifications/Compliance (Europe) CE, EN ISO 20345:2011, S2, SRC