Learn More
ABEBA™ X-Light White Occupational Shoes O2 ESD
Product Code. 17101515
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17101515 35 1 pair
17111515 36 1 pair
17101525 45 1 pair
17111525 46 1 pair
17121525 47 1 pair
17131525 48 1 pair
17121515 37 1 pair
17131515 38 1 pair
17141515 39 1 pair
17151515 40 1 pair
17161515 41 1 pair
17171515 42 1 pair
17181515 43 1 pair
17191515 44 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17101515 Supplier ABEBA™ Supplier No. 7131126/35
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
White X-light occupational shoes with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, PU slip resistant outsole
  • Lightweight, comfortable occupational shoes
  • Comply to EN ISO 20347: 2012, O2 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant Mono PU outsole SRC
  • White microfiber
  • Silver ions lining
  • Replaceable active comfort insole
Specifications
Color White
Material Microfiber
Size 35
Certifications/Compliance (Europe) Comply to EN 20347: 2012 O2 SRC, ESD dissipative according EN 61340
Gender Unisex
Ankle Style Low-cut
Waterproof Water Resistant
Product Type O2 SRC ESD