Learn More
Melford X-Phos BCIP
Product Code. 30018114
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Product Code. 30018114
Brand: Melford B740000.1
Colorimetric substrate for Alkaline phosphatase activity in blotting immunohistochemical and cytochemistry techniques.
Colorimetric substrate for Alkaline phosphatase activity in blotting immunohistochemical and cytochemistry techniques. Forms a purple insoluble precipitate when used in conjunction with NBT.
Specifications
Quantity 100 mg