Learn More
Draeger Safety X-Plore™ 4740 TPE Body
Product Code. 15704512
Change view
Click to view available options
:
Size: M/L
Product Code.
15704512 Size: M/L
1 options
Product Code. 15704512
Brand: Draeger Safety R55876
Only null left
Add to basket
Add to basket
Draeger Safety X-Plore™ 4740 TPE Body is a half mask that is robust but also offers excellent comfort and a tight fit to the face.
  • Designed to protect yourself from dangerous gases, vapors, and particles
  • Exhalation valve is positioned so moisture escapes the mask and a dry atmosphere is maintained
  • Hard mask body is made from durable PP plastic
  • Remove your mask without having to take off your safety hat using the drop-down strap system
Specifications
Certifications/Compliance CE certified (EN 140), AS/NZS
Size Medium/Large