missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xanthine Oxidase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-935-100ug
This item is not returnable.
View return policy
Description
Xanthine Oxidase Polyclonal antibody specifically detects Xanthine Oxidase in Bovine samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| Xanthine Oxidase | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| EC 1.17.1.4, EC 1.7.2.2, xanthene dehydrogenase, xanthine dehydrogenase, xanthine dehydrogenase/oxidase, xanthine oxidase, xanthine oxidoreductase, XDHA, XO, XOR | |
| Xanthine Oxidase [Bovine Buttermilk] (Uniprot: P80457) | |
| 100 μg | |
| Signal Transduction | |
| 7498 | |
| Bovine | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1:500-1:5000, ELISA 1:5000-1:20000, Immunoprecipitation 1:100 | |
| P80457 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Primary | |
| Reconstitute with 100 μg deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction