Learn More
SNV Comines™ Xavier™ Cotton Laboratory Coats
Product Code. 12671126
Change view
Click to view available options
Size:
40
44
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12671126 44 Each
12651126 40 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12671126 Supplier SNV Comines™ Supplier No. XAVLP00400T2
  • ONLINE EXCLUSIVE PRICE
    27.96€
    17.30€ / Pack of 1
    Save 10.66€
    38% Off
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides general light protection against splashes in the laboratory. SNV Comines™ Xavier™ Cotton Laboratory Coats are available in a range of sizes.
  • White in color
  • With snap buttons
  • Collared with lapels
Specifications
Product Type Xavier Lab Coat
Size 44
Gender Men's
Material Carbon
Color White
Closure Type Snaps
No. of Pockets 4
Size (Metric) Chest 96 cm
Includes 2 Flat pockets, 1 breast pocket, 1 inside pocket, opening angle
Construction 100% Cotton