Learn More
Waters Corporation XBridge Prep Shield RP18 Column
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development.
Brand:  Waters Corporation 186003997
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 13233310
  • 20400.00€ / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
Specifications
Specifications
30 mm
6000 psi (415 Bar)
10 μm
1/Ea.
Yes
250 mm
Spherical
130 Å
L1
Column