missing translation for 'onlineSavingsMsg'
Learn More

CELLPATH XMARK - TISSUE MARKING DYE

Product Code. 30226284
Change view
Click to view available options
Capacity:
60 mL
250 mL
Unit Size:
250mL
60mL
Product Code. Capacity unitSize
30226284 250 mL 250mL
30226283 60 mL 60mL
2 options
This item is not returnable. View return policy

Product Code. 30226284

Brand: CELLPATH RCA010100C

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

XMark Tissue Marking Dyes permanently mark surgical tissue margins in histological specimens. With nine colours in the range, each with a standard viscosity that ensures less variation from one colour to another and allows for easy marking on small or large specimens.

  • The optimised viscosity of the Xmark dye reduces viscosity on specimen application, achieving even dye coverage without dripping or running.
  • The high pigment concentrations in XMark dye derive highly durable marking and easy application to diagnostic specimens.
  • It coats the specimen tissue surface with minimal penetration. Its strong adhesion qualities mean the marking dye will not interfere with diagnostic interpretations, making them ideal for radical prostatectomies.
  • Available in nine colours, all easy to distinguish, reducing potential errors and ensuring the colour shown on the bottle (macroscopic colours) matches the colour observed under the microscope. Unlike other dyes XMark is non-hazardous, negating the need for special requirements during use.
  • Key Benefits:
  • Emulsified pigments are blended perfectly with an aqueous base to provide a consistent and high-quality product every time
  • Chemical and heat resistance to formalin, alcohols, xylene and wax provides clear visibility both before and after tissue processing
  • A 3-4 minute drying time improves laboratory workflow at the cut-up bench
  • Non-flammable

Specifications

Capacity 250 mL
Product Type Tissue Marking Dye
Solution Type Mordant Dye
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.