missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31083
This item is not returnable.
View return policy
Description
XPR1 Polyclonal specifically detects XPR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| XPR1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| XPR1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human XPR1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Xenopus laevis (93%) and Xenopus tropicalis (100%). Not yet tested in any other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| FLJ90308, Protein SYG1 homolog, SYG1xenotropic and polytropic retrovirus receptor, X3, Xenotropic and polytropic murine leukemia virus receptor X3, xenotropic and polytropic retrovirus receptor 1, XR, X-receptor | |
| Rabbit | |
| 82 kDa | |
| 100 μL | |
| GPCR, Signal Transduction | |
| 9213 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction