missing translation for 'onlineSavingsMsg'
Learn More

XRCC2 Mouse anti-Human, Clone: 3D9A1, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66652-1-IG-150UL

 View more versions of this product

Product Code. 16898725

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The x-ray repair cross-complementing (XRCC) proteins are responsible for efficiently repairing and maintaining genetic stability following DNA base damage. These genes share sequence similarity with the yeast DNA repair protein Rad5. XRCC2 and XRCC3 are both involved in maintaining chromosome stability during cell division. XRCC2 is required for efficient repair of DNA double-strand breaks by homologous recombination between sister chromatids, and XRCC3 interacts directly with Rad51 to cooperate with Rad51 during recombinational repair.
Specifications

Specifications

XRCC2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43543
XRCC2
XRCC2 Fusion Protein Ag14421
150 μL
Primary
Human
Antibody
IgG2b
Immunocytochemistry, Immunofluorescence, Western Blot
3D9A1
Unconjugated
XRCC2
DKFZp781P0919, DNA repair protein XRCC2, XRCC2
Mouse
Protein A
RUO
7516
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.