Learn More
XRCC2 Mouse, anti-Human, CoraLite 555, Clone: 3D9A1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL555-66652-20UL
 View more versions of this product
Product Code. 18005485
  • 145.00€ / 20µL
Description
The XRCC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human XRCC2. This antibody recognizes human, mouse antigen. The XRCC2 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™555.
Specifications
Specifications
XRCC2
3D9A1
CoraLite 555
Mouse
Primary
Human, Mouse
155 kDa
IgG2b
Flow Cytometry (Intracellular Staining)
1000 μg/mL
DKFZp781P0919, DNA repair protein XRCC2, XRCC2
20 μL
7516
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.