missing translation for 'onlineSavingsMsg'
Learn More

XRCC4 Mouse anti-Human, Clone: 3E5D2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66621-1-IG-150UL

 View more versions of this product

Product Code. 16878665

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The x-ray repair cross-complementing (XRCC) proteins are responsible for efficiently repairing and maintaining genetic stability following DNA base damage. These genes share sequence similarity with the yeast DNA repair protein Rad51. XRCC4 is an accessory factor of DNA ligase IV that preferentially binds DNA with nicks or broken ends. XRCC4 binds to DNA ligase IV and enhances its joining activity, and it is also involved in V(D)J recombination. Any defect in one of the known components of the DNA repair/V(D)J recombination machinery (Ku-70, Ku-80, DNA-PKCS, XRCC4 and DNA ligase IV) leads to abortion of the V(D)J rearrangement process and early block in both T and B cell maturation.
Specifications

Specifications

XRCC4
Monoclonal
0.83 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13426
XRCC4
XRCC4 Fusion Protein Ag8807
150 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
3E5D2
Unconjugated
XRCC4
DNA repair protein XRCC4, XRCC4
Mouse
Protein A
RUO
7518
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.