Learn More
XRN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11267-1-AP-150UL
 View more versions of this product
Product Code. 16878223
  • 410.00€ / 150µL
Description
XRN2 gene encodes a 5′-3′ exonuclease that promotes transcription termination at cotranscriptional cleavage sites. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
Specifications
XRN2
Polyclonal
Unconjugated
XRN2
5 3 exoribonuclease 2, 5′ 3′ exoribonuclease 2, DHM1 like protein, DHP protein, XRN2
Rabbit
Antigen Affinity Chromatography
RUO
22803, 24128, 362229
-20°C
Liquid
Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9DBR1, Q9H0D6
Xrn2
XRN2 Fusion Protein Ag1788
150 μL