missing translation for 'onlineSavingsMsg'
Learn More

Xylene Cyanol FF, 10g, MP Biomedicals

Product Code. 11442306 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442306 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11442306 Supplier MP Biomedicals Supplier No. 0521281710
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 2650-17-1
Molecular Formula C25H27N2NaO6S2
MDL Number MFCD00019481
InChI Key VVLFAAMTGMGYBS-KRQUPCAFSA-M
SMILES [Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O
IUPAC Name sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
Molecular Weight (g/mol) 538.61
Quantity 10 g
Chemical Name or Material Xylene Cyanol FF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.