missing translation for 'onlineSavingsMsg'
Learn More

Xylene Cyanol FF (Technical Grade), TRC

Product Code. 30449780
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449780 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449780 Supplier Toronto Research Chemicals Supplier No. TRCX74941310G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Xylene Cyanol FF
CAS 2650-17-1
Molecular Formula C25H27N2NaO6S2
Quantity 10 g
Synonym C.I. Acid Blue 147, C.I. Acid Blue 147','Monosodium Salt, Xylene Cyanole FF, Acid Blue 147, Acidine Pure Blue C Extra, Albion Xylene Cyanol, C.I. 42135, Cyanol Extra, Disulphine Blue FFN 200, Erio Glaucine FFX, Kiton Pure Blue FF, Merantine Blue FF, Xylene Blue FF, Xylene Cyanole
InChI Formula InChI=1S/C25H28N2O6S2.Na/c1-5-26-22-11-7-18(13-16(22)3)25(19-8-12-23(27-6-2)17(4)14-19)21-10-9-20(34(28,29)30)15-24(21)35(31,32)33;/h7-15,26H,5-6H2,1-4H3,(H,28,29,30)(H,31,32,33);/q;+1/p-1
SMILES O[C@H]1[C@@H](O)[C@@H](OP(OP(O[Na])(O[Na])=O)(O[Na])=O)O[C@@H]1COP(O[Na])(O[Na])=O
IUPAC Name sodium;4-[[4-(ethylamino)-3-methylphenyl]-(4-ethylimino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-3-sulfobenzenesulfonate
Molecular Weight (g/mol) 538.61
Formula Weight 538.1208
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.