Learn More
Thermo Scientific Chemicals Xylenol Orange, sodium salt, pure, water soluble
Product Code. 10174740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10174740 1 g Glass bottle 1g
10676802 5 g Glass bottle 5g
10421831 10 g Glass bottle 10g
11996931 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10174740 Supplier Thermo Scientific Chemicals Supplier No. 211220010
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
CAS: 3618-43-7 | C31H28N2Na4O13S
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3618-43-7
Molecular Formula C31H28N2Na4O13S
Specifications
Melting Point 210.0°C
Solubility Information Solubility in water: 510g/L (20°C).
Infrared Spectrum Authentic
Lambda Max 575 to 585nm (0.1N NaOH)
Formula Weight 760.59
Loss on Drying 10.0% (110°C, 1 hr) max.
Grade Pure
Packaging Glass bottle
Suitability (for Zinc titration) Passes test
UV Spectrometry Complies
Color Brown to Brown/Red
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Xylenol Orange, sodium salt