Learn More
YAP1 Antibody (CL13628) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24566AF350
Product Code. 30304336
  • 737.00€ / 0.10mL
Description
YAP1 Monoclonal antibody specifically detects YAP1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
YAP1
Monoclonal
Alexa Fluor 350
65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog
This antibody was developed using a synthetic peptide derived from P46937, with the exact immunogen sequence remaining proprietary.
0.1 mL
Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases
10413
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL13628
50mM Sodium Borate
Mouse
Protein A purified