missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ YIPF6 Recombinant Rabbit Monoclonal Antibody (6X1R7)

Rabbit Recombinant Monoclonal Antibody
176.00€ - 436.00€
Specifications
| Antigen | YIPF6 |
|---|---|
| Clone | 6X1R7 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284368
|
Invitrogen™
MA557883 |
20 μL |
176.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30284347
![]() |
Invitrogen™
MA557884 |
100 μL |
436.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: MAEAEDSPGE QEAAASKPLF AGLSDVSISQ DIPIEGEITI PSRARAQEHD SSTLNESIRR TIMRDLKAVG RKFMHVLYPR KSN.
YIPF6 is a multi-pass membrane proteinPotential. It belongs to the YIP1 family.The exact function of YIPF6 remains unknown.Specifications
| YIPF6 | |
| 1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q8BR70 | |
| 77929 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-83 of mouse Yipf6 (NP_997516.1). | |
| Antibody |
| 6X1R7 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| YIPF6 | |
| A430107J06Rik; FinGER6; Protein YIPF6; Yip1 domain family member 6; Yip1 domain family, member 6; YIP1 family member 6; yipf6; zgc:86904 | |
| YIPF6 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title