missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ YM 155

Product Code. 18798726
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18798726 10 mg 10mg
18705406 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18798726 Supplier R&D Systems™ Supplier No. 6491/10
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Survivin suppressor

YM 155 is a survivin suppressor. Down-regulates survivin expression and sensitizes human NSCLC cell lines to radiation. Enhances radiation-induced tumor regression in nude mice bearing NSCLC xenografts. Also induces apoptosis in TRAIL-resistant glioma cells in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,9-Dihydro-1-(2-methoxyethyl)-2-methyl-4,9-dioxo-3-(2-pyrazinylmethyl)-1H-naphth[2,3-d]imidazolium bromide
CAS 781661-94-7
Quantity 10 mg
Product Line Tocris
Molecular Formula C20H19BrN4O3
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.