missing translation for 'onlineSavingsMsg'
Learn More

YS-035 hydrochloride, Tocris Bioscience™

Product Code. 18023872
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18023872 10 mg 10mg
18085722 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18023872 Supplier Tocris Bioscience Supplier No. 0416/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Inhibits K+ outward/pacemaker current

A Ca2+ blocker that strikingly prolongs cardiac action potentials by inhibiting pacemaker current and K+ outward currents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89805-39-0
Molecular Formula C21H30ClNO4
Molecular Weight (g/mol) 395.924
InChI Key GYBONEHHSUABAO-UHFFFAOYSA-N
Synonym ys-035 hydrochloride, n,n-bis 2-3,4-dimethoxyphenyl ethyl methylamine hydrochloride, n,n-bis-3,4-dimethoxyphenylethyl-n-methylamine hydrochloride, acmc-20fbbm, bis 2-3,4-dimethoxyphenyl ethyl methyl amine hydrochloride, ys-035 hydrochloride, analytical standard, for drug analysis, 2-3,4-dimethoxyphenyl-n-2-3,4-dimethoxyphenyl ethyl-n-methylethanamine hydrochloride, n-2-3,4-dimethoxyphenyl ethyl-3,4-dimethoxy-n-methyl-benzeneethanamine hydrochloride, n--2-3,4-paragraph signthorn 1/4 xno+/-1/2 >>u oo>>u-3,4-paragraph signthorn 1/4 xno>>u-n-1/4 x>>u+/-1/2 oo degrees .
PubChem CID 6917841
IUPAC Name 2-(3,4-dimethoxyphenyl)-N-[2-(3,4-dimethoxyphenyl)ethyl]-N-methylethanamine;hydrochloride
SMILES CN(CCC1=CC(=C(C=C1)OC)OC)CCC2=CC(=C(C=C2)OC)OC.Cl

Specifications

Quantity 10 mg
Formula Weight 395.93
Percent Purity >98%
Chemical Name or Material YS-035 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.