Learn More
LNI Swissgas ZA TOTAL C 6.0
Product Code. 17841969 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage unitSize
17841969 100/115/220/230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17841969 Supplier LNI Swissgas Supplier No. 6920.25.360
Regulatory: This product is under eco-contribution tax.
Zero air generator for HC/carbon dioxide/moisture removal with integrated compressor.
  • Zero air generator 6.0 L/min HC/carbon dioxide/moisture removal with integrated compressor.
Specifications
Voltage 100/115/220/230 V
Type Zero Air Gas Generator