missing translation for 'onlineSavingsMsg'
Learn More

Zaleplon-D5, TRC

Product Code. 30453912
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453912 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30453912 Supplier Toronto Research Chemicals Supplier No. TRCZ1450021MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zaleplon-D5
CAS 1001083-56-2
Molecular Formula C17 2H5 H10 N5 O
Quantity 1 mg
Synonym Acetamide','N-[3-(3-cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-(ethyl-1,1,2,2,2-d5)- (ACI), N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-(ethyl-1,1,2,2,2-d5)acetamide (ACI), Zaleplon-d5, Zaleplon-D5 (N-ethyl-D5)
InChI Formula InChI=1S/C17H15N5O/c1-3-21(12(2)23)15-6-4-5-13(9-15)16-7-8-19-17-14(10-18)11-20-22(16)17/h4-9,11H,3H2,1-2H3/i1D3,3D2
SMILES [2H]C([2H])([2H])C([2H])([2H])N(C(=O)C)c1cccc(c1)c2ccnc3c(cnn23)C#N
IUPAC Name N-[3-(3-cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-(1,1,2,2,2-pentadeuterioethyl)acetamide
Molecular Weight (g/mol) 310.36
Formula Weight 310.159
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.