missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBED2 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00079413-B01P
This item is not returnable.
View return policy
Description
ZBED2 Polyclonal antibody specifically detects ZBED2 in Human samples. It is validated for Western Blot
Specifications
| ZBED2 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| MGC10796, zinc finger BED domain-containing protein 2, zinc finger, BED domain containing 2, zinc finger, BED-type containing 2 | |
| ZBED2 (NP_078784.2, 1 a.a. - 218 a.a.) full-length human protein. MMRREDEEEEGTMMKAKGDLEMKEEEEISETGELVGPFVSAMPTPMPHNKGTRFSEAWEYFHLAPARAGHHPNQYATCRLCGRQVSRGPGVNVGTTALWKHLKSMHREELEKSGHGQAGQRQDPRPHGPQLPTGIEGNWGRLLEQVGTMALWASQREKEVLRRERAVEWRERAVEKRERALEEVERAILEMKWKVRAEKEACQREKELPAAVHPFHFV | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| NP_078784.2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 79413 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction