missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€ - 589.00€
Specifications
| Antigen | ZBTB12 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18406561
|
Novus Biologicals
NBP2-33480-25ul |
25 μL |
415.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18151054
|
Novus Biologicals
NBP2-33480 |
0.1 mL |
589.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZBTB12 Polyclonal specifically detects ZBTB12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ZBTB12 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C6orf46, chromosome 6 open reading frame 46, D6S59E, G10Bat9, NG35HLA-B-associated transcript 9, Protein G10, zinc finger and BTB domain containing 12, zinc finger and BTB domain-containing protein 12 | |
| ZBTB12 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9Y330 | |
| 221527 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GLGIGGSVGGHLGELAQSSVPPSTVAPPQGVVKACY | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title