missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76518-0.025mg
This item is not returnable.
View return policy
Description
ZBTB5 Polyclonal specifically detects ZBTB5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| ZBTB5 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| KIAA0354, zinc finger and BTB domain containing 5, zinc finger and BTB domain-containing protein 5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9925 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| NP_055687 | |
| ZBTB5 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human ZBTB6. The immunogen is located within the last 50 amino acids of ZBTB5. Amino Acid Squence: KRFSQSSHLYKHSKTT | |
| 0.025 mg | |
| Primary | |
| At least two isoforms of ZBTB5 are known to exist; this antibody will recognize both isoforms. This antibody is predicted to not cross-react with other ZBTB protein family members. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction