missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H12D Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98171B
This item is not returnable.
View return policy
Description
ZC3H12D Polyclonal antibody specifically detects ZC3H12D in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ZC3H12D | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Biotin | |
| C6orf95, FLJ46041, MCP-induced protein 4, p34, TFL, transformed follicular lymphoma, tumor suppressor TFL, zinc finger CCCH-type containing 12D | |
| Produced in rabbits immunized with E. coli-derived Human ZC3H12D fragment. | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 340152 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering