missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCCHC17 Antibody (OTI9A7), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44773
This item is not returnable.
View return policy
Description
ZCCHC17 Monoclonal antibody specifically detects ZCCHC17 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| ZCCHC17 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| OTI9A7 | |
| Western Blot 1:2000 | |
| HSPC251, nucleolar protein of 40 kDa, Pnn-interacting nucleolar protein, pNO40nucleolar protein 40, PS1D protein, PS1DRP11-266K22.1, putative S1 RNA binding domain protein, Putative S1 RNA-binding domain protein, Zinc finger CCHC domain-containing protein 17, zinc finger, CCHC domain containing 17 | |
| Full length human recombinant protein of human ZCCHC17 (NP_057589) produced in E.coli. | |
| 100 μg | |
| metabolism | |
| 51538 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction