missing translation for 'onlineSavingsMsg'
Learn More

ZDHHC9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen ZDHHC9
Concentration 0.34 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16862635
View Documents
Proteintech
24046-1-AP-20UL
20 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
16852635
View Documents
Proteintech
24046-1-AP-150UL
150 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.
Specifications

Specifications

ZDHHC9
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Human
P59268, Q9Y397
208884, 51114
ZDHHC9 Fusion Protein Ag20881
Primary
-20°C
ZDHHC9
0.34 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CGI 89, CXorf11, DHHC 9, DHHC9, MMSA 1, Palmitoyltransferase ZDHHC9, UNQ261/PRO298, ZDHHC10, ZDHHC9, Zinc finger protein 379, ZNF379, ZNF380
ZDHHC9
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.