missing translation for 'onlineSavingsMsg'
Learn More

ZEB2 Monoclonal Antibody (OTI3D8), TrueMAB™, OriGene

Mouse Monoclonal Antibody

242.00€ - 540.87€

Specifications

Antigen ZEB2
Clone OTI3D8
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17540633
Encompass_Preferred
View Documents
Origene Technologies
TA802110S
30 μL
242.00€
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17520633
Encompass_Preferred
View Documents
Origene Technologies
TA802110
100 μL
540.87€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZEB2 Monoclonal antibody specifically detects ZEB2 in Human samples. It is validated for Western Blot
Specifications

Specifications

ZEB2
1 mg/mL
Monoclonal
Liquid
RUO
O60315
IgG1
-20° C, Avoid Freeze/Thaw Cycles
OTI3D8
Western Blot
Unconjugated
Mouse
Human
9839
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.