missing translation for 'onlineSavingsMsg'
Learn More

Zebrafish Basolateral Pole of Cells Antibody (FIS 2H9/1) - BSA Free, Novus Biologicals™

Product Code. 18615907 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18615907 0.025 mL 0.03mL
18633409 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18615907 Brand Novus Biologicals Brand No. NBP2503700.025ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Zebrafish Basolateral Pole of Cells Monoclonal antibody specifically detects Zebrafish Basolateral Pole of Cells in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zebrafish Basolateral Pole of Cells
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone FIS 2H9/1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Formulation PBS
Host Species Mouse
Immunogen Lysate of zebrafish intestine
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Developmental Biology, Epitope Tags
Primary or Secondary Primary
Target Species Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.