missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Gut Absorptive Cell Epitopes Antibody (FIS 4B7/1) - BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Zebrafish Gut Absorptive Cell Epitopes Monoclonal antibody specifically detects Zebrafish Gut Absorptive Cell Epitopes in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antigen | Zebrafish Gut Absorptive Cell Epitopes |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | FIS 4B7/1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Formulation | PBS |
| Host Species | Mouse |
| Immunogen | Zebrafish intestine |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?