missing translation for 'onlineSavingsMsg'
Learn More

Zebrafish Gut Secretory Cell Epitopes Antibody (FIS 4B7/2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

190.00€ - 351.00€

Specifications

Antigen Zebrafish Gut Secretory Cell Epitopes
Clone FIS 4B7/2
Concentration 1 mg/mL
Dilution Western Blot 1:200, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Applications Western Blot, Immunohistochemistry, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18614008
View Documents
Novus Biologicals
NBP2-50376-0.025ml
0.025 mL
190.00€
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18669127
View Documents
Novus Biologicals
NBP2-50376
0.1 mL
351.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zebrafish Gut Secretory Cell Epitopes Monoclonal antibody specifically detects Zebrafish Gut Secretory Cell Epitopes in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

Zebrafish Gut Secretory Cell Epitopes
1 mg/mL
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Mouse
Cell Cycle and Replication, Cellular Signaling, Developmental Biology, Epigenetics, Epitope Tags
PBS
IgG1
Protein A purified
FIS 4B7/2
Western Blot 1:200, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Monoclonal
Purified
RUO
Zebrafish
Lysate of fish intestine
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.