Learn More
Thermo Scientific™ Zeiss 20X Objective, Plan-Apochromat (0.8 NA)
Shop All Thermo Scientific ProductsDescription
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
Specifications
Specifications
| Type | Objective Lens |
| Product Line | Zeiss |
| Description | Zeiss 20X Objective, Plan-Apochromat (0.8 NA) |
| For Use With (Equipment) | ArrayScan HCA Readers |
| Lens Type | Dry |
| Magnification | 20x (0.8NA) |
| Quantity | 1 each |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
¿Detecta una oportunidad de mejora?