Learn More
Parker Hannifin™ Zero air generator 7.5L/min Domnick Hunter
Product Code. 12872706
Change view
Click to view available options
Flow Rate:
7.5 L/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate unitSize
12872706 7.5 L/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12872706 Supplier Parker Hannifin™ Supplier No. UHP75ZASE
  • ONLINE EXCLUSIVE PRICE
    16658.25€
    8803.64€ / Pack of 1
    Save 7854.61€
    47% Off
 Restricted Item
Estimated Shipment: 08-12-2026