Learn More
SI Analytics™ Zero Point Solution for Oxygen Electrodes
Product Code. 10788821 Shop All SI Analytics Products
Change view
Click to view available options
For Use With (Equipment):
Oxygen Electrodes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10788821 Oxygen Electrodes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10788821 Supplier SI Analytics™ Supplier No. 285138287
Add to basket
Add to basket
Suitable for oxygen electrodes, models OX 1100 and OX 1100 +
  • For oxygen electrodes OX 1100/OX 1101
  • Supplied in ampoules
TRUSTED_SUSTAINABILITY
Specifications
Description Supplied in 60 Ampules
For Use With (Equipment) Oxygen Electrodes