Learn More
Zeta Opioid Receptor Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94528-0.02ml
This item is not returnable.
View return policy
Description
Zeta Opioid Receptor Polyclonal antibody specifically detects Zeta Opioid Receptor in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Zeta Opioid Receptor | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| 7-60, 7-60 protein, OGFr, opioid growth factor receptor, Protein 7-60, Zeta-type opioid receptor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human Zeta Opioid Receptor (NP_031372.2). MDDPDCDSTWEEDEEDAEDAEDEDCEDGEAAGARDADAGDEDEESEEPRAARPSSFQSRMTGSRNWRATRDMCRYRHNYPDLVERDCNGDTPNLSFYRNEIRFLPNGCFIEDILQNWTDNYDLLEDNHSYIQWLFPLREPGVNWHAKPLTLREVEVFKSSQEIQERLVRAYELMLGFYGIRLEDRGTGTVGRAQNYQKRFQNLNWRSHNNLRITRILKSLGELGLEHFQAPLVRFFLEETLVRRELPGVRQSALDYFMFAVRCRHQRRQLVHFAWEHFRP | |
| 0.02 mL | |
| Cell Biology, Cell Cycle and Replication, GPCR, Signal Transduction | |
| 11054 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |