missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP36L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12306-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a member of the TIS11 family of early response genes. Family members are induced by various agonists such as the phorbol ester TPA and the polypeptide mitogen EGF. The gene is well conserved across species and has a promoter that contains motifs seen in other early-response genes. The encoded protein contains a distinguishing putative zinc finger domain with a repeating cys-his motif. This putative nuclear transcription factor most likely functions in regulating the response to growth factors.Specifications
| ZFP36L1 | |
| Polyclonal | |
| Unconjugated | |
| ZFP36L1 | |
| Berg36, BRF1, Butyrate response factor 1, cMG1, EGF response factor 1, ERF 1, ERF1, Protein TIS11B, RNF162B, TIS11B, ZFP36L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12192, 29344, 677 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P17431, P23950, Q07352 | |
| ZFP36L1 | |
| ZFP36L1 Fusion Protein Ag2952 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction