Learn More
ZFP37 Monoclonal Antibody (OTI4C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810216S
Description
ZFP37 Monoclonal antibody specifically detects ZFP37 in Human samples. It is validated for Western BlotSpecifications
| ZFP37 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P60510 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5531 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4C2 | |
| Unconjugated | |
| PPP4C | |
| LOW QUALITY PROTEIN: zinc finger protein 37 homolog; male germ cell-specific zinc finger protein; Tzn; ZFP37; Zfp-37; ZFP37 zinc finger protein; zinc finger protein 37; Zinc finger protein 37 homolog; zinc finger protein homologous to Zfp37 in mouse; ZNF906 | |
| Full length human recombinant protein of human PPP4C produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.