missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP90 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540402
This item is not returnable.
View return policy
Description
Peptide sequence: QEECGGAQSL GNCDQVLRGY QVSKPEVIFK LEQGEEPWIS EGEIQRPFYP.
ZFP90 may function as a repressor or silencer protein, and most likely exerts its repressing activity upon zinc-dependent binding to DNA. It may be involved in proper spermatogenesis by repressing the expression of genes unnecessary or incompatible with the maintenance of a haploid cell state.
Specifications
| ZFP90 | |
| Polyclonal | |
| Unconjugated | |
| ZFP90 | |
| 6430515L01Rik; FIK; FOXP3-interacting KRAB domain-containing protein; KIAA1954; KRAB17; mKIAA1954; NK10; Nk10 expressed protein; Zfp64; Zfp83; Zfp90; zfp-90; ZFP90 zinc finger protein; zinc finger protein 476; zinc finger protein 756; zinc finger protein 83; zinc finger protein 90; Zinc finger protein 90 homolog; zinc finger protein NK10; ZNF756 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 146198 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TF47 | |
| ZFP90 | |
| Synthetic peptide directed towards the middle region of human ZFP90. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction