missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika virus Capsid Protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-13200
This item is not returnable.
View return policy
Description
Zika virus Capsid Protein Polyclonal antibody specifically detects Zika virus Capsid Protein in Virus samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Zika virus Capsid Protein | |
| Polyclonal | |
| Western Blot 1:500-1:20000, Immunofluorescence 1:100-1:1000 | |
| Capsid Protein, Capsid protein C, Core protein, ZIKV Capsid protein | |
| Full length Zika virus Capsid recombinant protein. (Zika virus (strain H/PF/2013)) | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction