Learn More
Zika Virus (SPH2015) NS1 Antibody (15), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-05871-50ul
 View more versions of this product
Product Code. 18643926
  • 265.00€ / 50µL
Description
Zika Virus (SPH2015) NS1 Monoclonal antibody specifically detects Zika Virus (SPH2015) NS1 in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Zika Virus (SPH2015) NS1
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Mouse
Protein A purified
RUO
Virus
Purified
Western Blot, ELISA
15
Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000
Zika NS1, ZIKV NS1, ZIKV-NS1
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Zika Virus (Accession#: ALU33341.1; Val796-Leu1157).
50 μg
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.