missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Zinc Reagent Sets, For Colorimetric Water Testing

Product Code. 11773283
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11773283 1 Set 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11773283 Supplier Hanna Instruments™ Supplier No. HI9373101

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Zinc Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.

These high quality reagents are designed to follow Standard Methods for the Examination of Water and Wastewater, 20th edition, Zincon method for colorimetric determination. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.

  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability

Specifications

Analytes Zincon
For Use With (Application) Zinc determination
For Use With (Equipment) Compatible Hanna benchtop and portable photometers
Includes Dropper bottle and individual packets
No. of Tests 100
Quantity 1 Set
Range 0.00 to 3.00mg/L zinc
Test Method Cholorimetric determination
Type Water Testing Kit
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H332-Harmful if inhaled.
  • H360FD-May damage fertility. May damage the unborn child.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.
  • P370+P378-In case of fire: Use to extinguish.
  • P391-Collect spillage.
  • P404-Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contain: potassium tetraborate, diboron trioxide,potassium cyanide,cyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.