missing translation for 'onlineSavingsMsg'
Learn More

Zinc sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.99% (metals basis), Thermo Scientific™

Product Code. 15474865
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474865 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15474865 Supplier Thermo Scientific Alfa Aesar Supplier No. 041658.KS
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-66-6
Quantity 1 Ea.
Odor Odorless
Molecular Formula Zn
MDL Number MFCD00011291
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
Solubility Information Soluble in water.
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES [Zn]
IUPAC Name zinc
Molecular Weight (g/mol) 65.38
PubChem CID 23994
ChEBI CHEBI:30185
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Zinc sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.