missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate Heptahydrate, TRC

Product Code. 30449944
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449944 250 mg -
30450595 2.5 g 2.5g
30454700 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30449944 Supplier Toronto Research Chemicals Supplier No. TRCZ701633250MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-20-0
Molecular Formula O4 S . Zn . 7 H2 O
Molecular Weight (g/mol) 287.58
Synonym Sulfuric acid, zinc salt (1:1), heptahydrate (8CI,9CI), White vitriol, Zinc sulfate (ZnSO4) heptahydrate, Zinc sulfate heptahydrate, Zinc vitriol, Zinc(II) sulfate heptahydrate
IUPAC Name zinc;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Zn+2].[O-]S(=O)(=O)[O-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2
Formula Weight 285.9548
Chemical Name or Material Zinc sulfate heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.