missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate Solution, 0.1M, Eur. Ph. Grade, Chem-Lab

Product Code. 12608533
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12608533 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12608533 Supplier Chem-Lab Supplier No. CL07.2618.1000
  • ONLINE EXCLUSIVE PRICE
    102.00€
    51.50€ / 1 L
    Save 50.50€
    49% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured according to Eur. Ph. 4(4.2.2) standards. Zinc Sulfate Solution, 0.1M, Eur. Ph. Grade, Lab-Chem provides a source of zinc for titrations in laboratory applications, including in volumetric solutions that that have been standardized in EDTA.

Zinc Sulfate Heptahydrate is a moderately water and acid soluble Zinc source for uses compatible with sulfates. This solution contains 28.754g per liter (0.1mol/liter).

Specifications

CAS 7446-20-0, 7732-18-5
Color Colorless
CAS Max % 3%
Physical Form Liquid
Quantity 1 L
CAS Min % 97%
Odor Odorless
Density 1.01g/mL
Molecular Formula H14O11SZn
Linear Formula H14O11SZn
MDL Number MFCD00149894
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
IUPAC Name zinc;sulfate;heptahydrate
Molecular Weight (g/mol) 287.54
PubChem CID 62640
ChEBI CHEBI:32312
Formula Weight 287.54g/mol
Grade Eu. Ph. Chapter 4(4.2.2)
Concentration or Composition (by Analyte or Components) 0.1mol/L (±0.0002 at 20°C)
Chemical Name or Material Zinc sulfate heptahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Zinc sulfate 0.1 mol/l
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: zinc sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.