Learn More
Zinquin Ethyl Ester, TRC
Product Code. 30457219
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457219 1 mg -
30455689 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30457219 Supplier Toronto Research Chemicals Supplier No. TRCZ4500001MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 181530-09-6
Molecular Formula C21 H22 N2 O5 S
Molecular Weight (g/mol) 414.47
Synonym 2-[[2-Methyl-8-[[(4-methylphenyl)sulfonyl]amino]-6-quinolinyl]oxy]acetic Acid, Zinquin Zinquin A, [[2-Methyl-8-[[(4-methylphenyl)sulfonyl]amino]-6-quinolinyl]oxy]-acetic Acid
IUPAC Name ethyl 2-[2-methyl-8-[(4-methylphenyl)sulfonylamino]quinolin-6-yl]oxyacetate
SMILES CCOC(=O)COc1cc(NS(=O)(=O)c2ccc(C)cc2)c3nc(C)ccc3c1
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C21H22N2O5S/c1-4-27-20(24)13-28-17-11-16-8-7-15(3)22-21(16)19(12-17)23-29(25,26)18-9-5-14(2)6-10-18/h5-12,23H,4,13H2,1-3H3
Formula Weight 414.1249
Chemical Name or Material Zinquin Ethyl Ester