missing translation for 'onlineSavingsMsg'
Learn More

Zinterol hydrochloride, Tocris Bioscience™

Product Code. 18772794
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18772794 10 mg 10mg
18712584 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18772794 Supplier Tocris Bioscience Supplier No. 1051/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective β2 agonist

Potent and selective β2-adrenoceptor agonist (pKB values are 8.3 and < 5.7 for β 2 and β1 receptors respectively, as measured in human artery).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37000-20-7
Molecular Formula C19H26N2O4S
Molecular Weight (g/mol) 378.487
InChI Key XJBCFFLVLOPYBV-UHFFFAOYSA-N
Synonym zinterol, zinterol inn, zinterolum inn-latin, n-2-hydroxy-5-1-hydroxy-2-2-methyl-1-phenylpropan-2-ylamino ethyl phenyl methanesulfonamide, n-2-hydroxy-5-1-hydroxy-2-2-methyl-1-phenylpropan-2-yl amino ethyl phenyl methanesulfonamide, zinterolum, 2'-hydroxy-5'-l-hydroxy-2-2-methyl-1-phenyl-2-propylamino ethyl methanesulfonanilide, methanesulfonamide, n-5-2-1,1-dimethyl-2-phenylethyl amino-1-hydroxyethyl-2-hydroxyphenyl, d0a0kp
PubChem CID 37990
IUPAC Name N-[2-hydroxy-5-[1-hydroxy-2-[(2-methyl-1-phenylpropan-2-yl)amino]ethyl]phenyl]methanesulfonamide
SMILES CC(C)(CC1=CC=CC=C1)NCC(C2=CC(=C(C=C2)O)NS(=O)(=O)C)O

Specifications

Quantity 10 mg
Formula Weight 414.95
Percent Purity >98%
Chemical Name or Material Zinterol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.