Learn More
Greiner Bio-One Zip Bags
Product Code. 12509146
Change view
Click to view available options
Dimensions (L x W):
80 x 120mm
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12509146 80 x 120mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12509146 Supplier Greiner Bio-One Supplier No. SSPA100
  • ONLINE EXCLUSIVE PRICE
    196.99€
    93.42€ / Pack of 1000
    Save 103.57€
    52% Off
View all promotions
Estimated Shipment: 21-10-2026
Specifications
Closure Type Zip Seal
Quantity 1000 Pk.
Dimensions (L x W) 80 x 120mm
Product Type Zip-seal Storage Bag